HPLC Analysis of Various Polymers on TSKgel® SuperMultiporeHZ-H
HPLC Analysis of Various Polymers on TSKgel® SuperµLtiporeHZ-H
Conditions
column TSKgel SuperµLtiporeHZ-N, 15 cm x 4.6 mm I.D., 6 µm particles (821885)
column temp. 40 °C
mobile phase THF
flow rate 0.35 mL/min
sample each polymer, 3 mg/mL
injection 10 µL
detector refractive index (RI)
Description
Categories Analytical Chromatography, Polymers, coatings, and plastics
Featured Industry Chemicals and Industrial Polymers
Legal Information TSKgel is a registered trademark of Tosoh Corporation
Other Notes Data provided by Tosoh Bioscience LLC.