HPLC Analysis of Vitamins, Fat Soluble (D2 and D3), on Discovery® HS C18
HPLC Analysis of Vitamins, Fat Soluble (D2 and D3), on Discovery® HS C18
Conditions
column Discovery HS C18, 15 cm x 4.6 mm I.D., 3 µm particles (569252-U)
column temp. 30 °C
mobile phase 100% acetonitrile
flow rate 0.8 mL/min
sample 50 µg/mL each analyte
injection 10 µL
detector UV, 290 nm
Description
Categories Analytical Chromatography, Vitamins and Natural products
Featured Industry Clinical
Vitamins, Nutraceuticals, and Natural Products
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC