HPLC Analysis of Warfarin™ Enantiomers on Astec® CHIROBIOTIC® V (Method 1)
HPLC Analysis of Warfarin™ Enantiomers on Astec® CHIROBIOTIC® V (Method 1)
Conditions
column CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 µm particles (11024AST)
column temp. 25 °C
mobile phase [A] acetonitrile; [B] 5 mM ammonium acetate, pH 4.1; (30:70, A:B)
flow rate 1 mL/min
sample 1 mg/mL in mobile phase
injection 5 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Clinical, anticoagulants, Clinical, Forensic & Toxicology, Anticoagulants, Pharmaceutical (small molecule), Anticoagulants/Antiplatelet Drugs, All Pharmaceutical Applications, Therapeutic Drug Monitoring
Featured Industry Clinical
Pharmaceutical (small molecule)
Legal Information Warfarin is a trademark of Wisconsin Alumni Research Foundation, CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC