HPLC Analysis of Water Soluble Vitamins on Ascentis® RP-Amide
HPLC Analysis of Water Soluble Vitamins on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 30 °C
mobile phase 25 mM potassium phosphate monobasic, pH 3.5
flow rate 1.0 mL/min
injection 10 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Vitamins and Natural products, Anticonvulsant/antiepiletic/antiseizure
Featured Industry Vitamins, Nutraceuticals, and Natural Products