HPLC Analysis of Xylene Isomers on Discovery® Zr-Carbon
HPLC Analysis of Xylene Isomers on Discovery® Zr-Carbon
Conditions
column Discovery Zr-Carbon, 15 cm x 4.6 mm I.D., 3 µm particles (65730-U)
column temp. 30 °C
mobile phase [A] water: [B] acetonitrile; (50:50, A:B)
flow rate 1 mL/min
sample o-xylene, m-xylene, p-xylene (290 µg/mL)
injection 1 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Semivolatiles
Featured Industry Chemicals and Industrial Polymers
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC