Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis Proteins on Proteomix® SCX-NP1.7, Demonstration of Column Stability with Repeated Injections
HPLC Analysis Proteins on Proteomix® SCX-NP1.7, Demonstration of Column Stability with Repeated Injections
Conditions
column Proteomix SCX-NP1.7, 5 cm x 4.6 mm I.D., 1.7 µm particles (Z777146)
column Proteomix SCX-NP1.7 guard cartridge with holder, 1 cm x 4 mm I.D. (Z777144)
column temp. ambient
mobile phase (A) 20 mM sodium phosphate buffer, pH 6.5, (B) A + 1.0 M NaCl
gradient 0 - 100% B in 25 min, 15 min prewash
flow rate 0.5 ml/min
sample each compound 1 mg/mL
injection 5 µl
detector UV, 214 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Proteomix is a registered trademark of Sepax Technologies