IC Separation of Chloride, Nitrite, Bromide, Nitrate, Phosphate, and Sulfate Ions on Metrohm Metrosep A Supp 10
IC Separation of Chloride, Nitrite, Bromide, Nitrate, Phosphate, and Sulfate Ions on Metrohm Metrosep A Supp 10
Conditions
column Metrosep A Supp 10, 10 cm x 4 mm I.D., 5 µm particles (Metrohm 6.1020.010)
column temp. 45 °C
mobile phase [A] 5.0 mM sodium carbonate; [B] 5.0 mM sodium bicarbonate
flow rate 1 mL/min
sample chloride (2 mg/L), nitrite (5 mg/L), bromide, nitrate, phosphate, sulfate (10 mg/L)
injection 20 µL
detector conductivity
Description
Categories Analytical Chromatography, Anions, Ion chromatography
Featured Industry Chemicals and Industrial Polymers
Environmental
Food and Beverages
General Analytical