LC/MS Analysis of 2C Amines Drugs of Abuse on Ascentis® Express HILIC
LC/MS Analysis of 2C Amines Drugs of Abuse on Ascentis® Express HILIC
Conditions
column Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 µm particles (53939-U)
column temp. 35 °C
mobile phase 5 mM ammonium formate in acetonitrile:water (90:10, v/v)
flow rate 0.4 mL/min
sample 300 ng/mL in acetonitrile
detector 2.0 µL
detector TOF/MS
Description
Categories Analytical Chromatography, Clinical, drugs of abuse, Drugs of Abuse
Featured Industry Clinical
Forensics and Toxicology