LC/MS Analysis of Anti-infectives on Ascentis® Express Biphenyl
LC/MS Analysis of Anti-infectives on Ascentis® Express Biphenyl
Conditions
column Ascentis Express Biphenyl, 10 cm x 3.0 mm, 2.7 µm particles (64066-U)
column temp. 35 °C
gradient 10 mM ammonium formate (70:30 water:acetonitrile) SW pH 4.00
flow rate 0.6 mL/min
pressure 997 psi (68.7 bar)
sample 300ng/mL in 85:15,  water:methanol
injection 5 µL
detector MS, ESI+, XIC
Description
Featured Industry Cosmetics, Personal Care, and Cleaning Products
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for LC-MS