LC/MS Analysis of Methadone and Metabolites EDDP and EMDP on Ascentis® Express RP Amide
LC/MS Analysis of Methadone and Metabolites EDDP and EMDP on Ascentis® Express RP Amide
Conditions
column Ascentis Express RP Amide, 10 cm x 2.1 mm I.D., 2.7 µm particles (53913-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium formate in water, pH 3.6; [B] 10 mM ammonium formate in acetonitrile, pH 3.6; (65:35, A:B)
flow rate 0.5 mL/min
sample 200 µg/L in 25:75, water: acetonitrile
injection 0.5 µL
detector ESI(+), m/z 100-1000
Description
Categories Analytical Chromatography, Forensics and Toxicology, analgesics, Drugs of Abuse, drugs of abuse
Featured Industry Forensics and Toxicology