LC/MS Analysis of Methamphetamine Enantiomers on Astec® CHIROBIOTIC® V2
LC/MS Analysis of Methamphetamine Enantiomers on Astec® CHIROBIOTIC® V2
Conditions
column Chirobiotic V2  15 cm x 4.6 mm, 5 µm particle size (15023AST)
column temp. 20 °C
mobile phase [A] methanol; [B] water; [C] acetic acid; [D] ammonium hydroxide; (95:5:0.1:0.02, A:B:C:D)
flow rate 1.0 mL/min
pressure 1220 psi (84 bar)
sample (+/-)-Methamphetamine 100 ng/mL in methanol
injection 2 µL
detector MS, ESI(+),SIR m/z 150.1
Description
Categories Analytical Chromatography, Forensics and Toxicology, Drugs of Abuse, Enantiomers, enantiomers, drugs of abuse
Featured Industry Forensics and Toxicology
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for LC-MS