LC/MS Analysis of Omeprazole and Metabolites on Ascentis® Express RP Amide (pH 2.7)
LC/MS Analysis of Omeprazole and Metabolites on Ascentis® Express RP Amide (pH 2.7)
Conditions
column Ascentis Express RP-Amide, 5 cm x 2.1 mm I.D., 2.7 µm particles (53911-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium formate in water, pH 2.7; [B] 10 mM ammonium formate in methanol, pH 2.7; (65:35, A:B)
flow rate 0.3 mL/min
sample 1 mg/L in mobile phase
injection 1 µL
detector ESI(+), m/z 50-400
Description
Analysis Note This application demonstrates the suitability of the Ascentis Express RP Amide for the analysis of omeprazole metabolites.
Categories Analytical Chromatography, Clinical, antiulcer compounds, Antiulcer Compounds