LC/MS Analysis of Polypeptide Drugs on Ascentis® Express RP-Amide
LC/MS Analysis of Polypeptide Drugs on Ascentis® Express RP-Amide
Conditions
column Ascentis Express RP-Amide, 10 cm x 2.1 mm I.D., 2.7 µm particles (53913-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium acetate in water, pH 4.2; [B] 10 mM ammonium acetate in 90:10 (v/v) acetonitrile:water, pH 4.7
gradient held at 20% B for 0.1 min; 20 to 80% B in 2.9 min; 80 to 20% B in 0.01 min; held at 20% B for 0.99 min
flow rate 0.3 mL/min
sample 1 µL/mL in 10mM ammonium formate, 80:20, acetonitrile:water
injection 3 µL
detector TOF, ESI(+), m/z 100-2500
Description
Analysis Note This application demonstrates the suitability of the Ascentis Express RP Amide for the analysis of polypeptide drugs.
Categories Analytical Chromatography, Peptides