LC/MS Analysis of Triglycerides in Soybean Oil on Ascentis® C18
LC/MS Analysis of Triglycerides in Soybean Oil on Ascentis® C18
Conditions
column two Ascentis C18, 15 cm x 2.1 mm I.D., 3 µm particles connected in series (581302-U)
column temp. 25 °C
mobile phase [A] 2-propanol: [B] acetonitrile; (60:40, A:B)
flow rate 0.2 mL/min
sample soybean oil weighed and diluted in acetone (10 mg in 1 ml w/v)
injection 2 µL
detector MS APCI mode
Description
Categories Analytical Chromatography, Fatty Acids, FAMEs