LC/MS Analysis of Zopiclone and Metabolites in Urine on Ascentis® Express ES-Cyano 2.0 μm
LC/MS Analysis of Zopiclone and Metabolites in Urine on Ascentis® Express ES-Cyano 2.0 μm
Conditions
sample preparation combine 100 µL urine spiked with zopiclone and metabolites at 10 ng/mL, 100 µL deuterated internal standards at 100 ng/mL in acetonitrile, and 800 µL water. centrifuge at 3000 rcf for 1 min.
column Ascentis Express ES-Cyano, 10 cm x 3.0 mm I.D., 2.0 µm particles (51732-U)
column temp. 25 °C
mobile phase [A] 10 mM ammonium formate pH 3.0 with formic acid; [B] acetonitrile; (70:30, A:B)
flow rate 0.4 mL/min
pressure 5300 psi (365 bar)
sample 1 ng/mL in urine:water:acetonitrile (10:80:10)
injection 10 µL
detector MS, MRM, m/z 375/245, 389/245, 405/245
Description
Analysis Note Zopiclone is a nonbenzodiazepine hypnotic agent used in the treatment of insomnia. It is one of the so-called "Z-drugs." Because they are additive and can be abused, their analysis in urine is necessary. Shown here is the rapid, sensitive analysis of zopiclone and its metabolites from urine on a Fused-Core Ascentis Express column under UHPLC conditions. Cerilliant CRMs provided reliable quantification.
Categories Analytical Chromatography, Drugs of Abuse
Featured Industry Clinical
Forensics and Toxicology
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for LC-MS