LC/MS (TOF) Analysis of Malvinidin Glycosides from High Bush Blueberries on Ascentis® Express C18
LC/MS (TOF) Analysis of Malvinidin Glycosides from High Bush Blueberries on Ascentis® Express C18
Conditions
column Ascentis Express C18, 10 cm x 2.1 mm I.D., 2.7 µm particles (53823-U)
column temp. 35 °C
mobile phase [A] 0.1% (v/v) formic acid in water; [B] 0.1% (v/v) formic acid in 75:25 (v/v) acetonitrile:water
gradient 2% B for 2 min, to 100% B in 38 min
flow rate 0.2 mL/min
injection 1 µL
detector ESI(+) TOF, extracted ions m/z 493.1354, 463.1244, 535.1481
Description
Analysis Note The purpose of this study was to measure the change in anthocyanins levels in wild blueberries as a function of berry development. The delphinidin, putinidin, peonidin, and malvinidin compounds exhibited a dramatic increase in abundance as a function of berry development. The cyanidin compounds exhibited a much lower increase. (The significance of the rate of increase is uncertain.) Chlorogenic acid exhibited a continuous decrease in abundance as a function of berry development. Overall, the combination of UV/Vis detection along with accurate mass TOF enabled an effective method of identification of anthocyanins from blueberry extracts. The Ascentis Express column provided rapid, sensitive detection and rugged operation.
Categories Analytical Chromatography, Antioxidants
Featured Industry Food and Beverages
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Berries (1.0 g) were added to 1.0 mL of 1% (v/v) formic acid in methanol. Samples were crushed in the solvent mixture and extracted (refrigerated) for 2 hours. A portion of the extracted sample was removed, centrifuged and the supernatant was collected for HPLC analysis.
suitability application for LC-MS