HPLC Analysis of Inhibin α-Subunit Fragment 1-32 on Discovery® HS F5
HPLC Analysis of Inhibin α-Subunit Fragment 1-32 on Discovery® HS F5
Conditions
column Discovery HS F5, 10 cm x 2.1 mm ID, 5 µm particles (567510-U)
column temp. 22 °C
mobile phase [A] 80 mM formic acid, 40 mM ammonium hydrate (pH ~3.5) in water; [B] 80 mM formic acid, 40 mM ammonium hydrate in 80% acetonitrile
flow rate 0.2 mL/min
sample 50 mg/L in 0.1 % formic acid
injection 2 µL
detector ESI (+), full scan
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC