Stability and Reproducibility of Zenix® 300 under High Salt and High pH Mobile Phase Conditions
Stability and Reproducibility of Zenix® 300 under High Salt and High pH Mobile Phase Conditions
Conditions
column Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 µm particles (Z777018)
column temp. 25 °C
mobile phase 50 mM sodium phosphate + 1 M NaCl, pH 8.5
flow rate 1.0 mL/min
pressure 1450 psi (100 bar)
sample each compound 1 mg/mL except uracil, 0.1 mg/mL
injection 5 µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information Zenix is a registered trademark of Sepax Technologies