Typical Desalting Chromatograms on TSKgel® BioAssist® DS HPLC Columns
Typical Desalting Chromatograms on TSKgel® BioAssist® DS HPLC Columns
Conditions
column TSKgel BioAssist DS, 15 cm x 4.6 mm I.D., 13 µm particles (821463)
column temp. ambient
mobile phase 10 mM phosphate buffer, 0.1 M KH2PO4/K2HPO4, pH 6.7, 10 mM Na2SO4, 0.005% sodium azide
flow rate 1 mL/min
sample gamma-globulin peak fraction
injection 10 µL
detector UV, 280 nm and refractive index (RI)
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information BioAssist is a registered trademark of Tosoh Corporation, TSKgel is a registered trademark of Tosoh Corporation
Other Notes Data provided by Tosoh Bioscience LLC.