UHPLC Analysis of Fat Soluble Vitamins on Titan™ C18, Methanol Mobile Phase
UHPLC Analysis of Fat Soluble Vitamins on Titan™ C18, Methanol Mobile Phase
Conditions
column Titan C18, 50 mm x 2.1 mm I.D., 1.9 µm particles (577122-U)
column temp. 30 °C
mobile phase 100% methanol
flow rate 0.4 mL/min
pressure 2741 psi (189 bar)
sample All compounds at 0.5 mg/mL except retinyl acetate at 0.05 mg/mL in methanol
injection 0.5 µL
injection UV, 280 nm
Description
Categories Analytical Chromatography, Vitamins and Natural products
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC