UHPLC Analysis of Kathon on Titan™ C18 using MS Detection
UHPLC Analysis of Kathon on Titan™ C18 using MS Detection
Conditions
column Titan C18 50 mm x 2.1 mm, I.D, 1.9 µm particles (577122-U)
mobile phase [A] 0.1% formic acid in water [B]:methanol
flow rate 500 µL/min
sample Diluted Kathon at 1 µg/mL in 95:5 0.1% formic acid in water::methanol
injection 1 µL
detector MS, ESI (+) MRM TIC
detector Diode Array at 275 nm
Description
Categories Analytical Chromatography, Disinfectants, Preservatives, Antiseptic/disinfectants
Featured Industry Cosmetics, Personal Care, and Cleaning Products
Environmental
General Analytical
Pharmaceutical (small molecule)
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC