UHPLC Analysis of p-Acetophenetide (Phenacetin) Titan™ C18
UHPLC Analysis of p-Acetophenetide (Phenacetin) Titan™ C18
Conditions
column TITAN C18, 10 cm x 2. 1 mm I.D., 1.9 um particles (577124-U)
column temp. 35 °C
mobile phase (A) 10 mM ammonium acetate, pH 7.0; (B) acetonitrile (70:30, A;B)
flow rate 0.3 mL/min
sample 300 ng/mL each in acetonitrile:water (3:1)
injection 2 µL
detector MS, XIC m/z 180.10
Description
Categories Analytical Chromatography, Analgesics
Featured Industry Pharmaceutical (small molecule)
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC