UHPLC Analysis of Turmeric Standards on Ascentis® Express RP-Amide
UHPLC Analysis of Turmeric Standards on Ascentis® Express RP-Amide
Conditions
column Ascentis Express RP-Amide, 10 cm x 4.6 mm I.D., 5 µm particles (50774-U)
column temp. 35 °C
mobile phase [A] water with 0.1% formic acid; [B] acetonitrile with 0.1% formic acid;
gradient 40% B held for 1 min; 40 to 100% B in (6 min); 100% B held for 5 min
flow rate 1.0 mL/min
pressure 1750 psi (121 bar)
sample 7 turmeric standards 50 µg/mL in 50:50, water:methanol
injection 5 µL
detector UV, 200 nm
Description
Categories Analytical Chromatography, Flavors, Fragrances, Herbal medicines
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC