UHPLC-MS/MS Analysis of Multiple Mycotoxins on Titan™ C18
UHPLC-MS/MS Analysis of µLtiple Mycotoxins on Titan™ C18
Conditions
column Titan C18, 10 cm x 2.1 mm I.D., 1.9 µm particles (577124-U)
column temp. 40 °C
mobile phase [A] 5mm ammonium formate with 0.1% formic acid in water ; [B] 5mm ammonium formate with 0.1% formic acid in acetonitrile
gradient 30% to 60%B in 3 min, to 100%B in 2 min, hold at 100%B for 2 min, 100%B to 30%B in 0.5 min, hold at 30% for 2 min
flow rate 0.4 mL/min
pressure 8557 psi (590 bar)
injection 2 µL
detector MS/MS, ESI(+), MRM
Description
Analysis Note Titan C18 UHPLC column provided rapid resoltion of mulitple mycotoxins.
Categories Analytical Chromatography, Mycotoxins
Featured Industry Clinical
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC-MS