Analytical Method: Analytical Quality Assurance, Standard for Anionic Surfactants
Reagents:
Cat.No. 112146  Dodecane-1-sulfonic acid salt for tenside tests
Cat.No. 116754  Water for analyis
Preparation of a standard solution:
Dissolve 1.00 g of sodium dodecane-1-sulfonis acid with distilled water in a calibrated or conformity-checked
1000-ml volumetric flask and make up to the mark with distilled water. The stock solution prepared according to this procedure has a concentration of 1000 mg/l anionic surfactants.
Further investigational concentrations may be prepared from this stock solution by diluting accordingly.
Stability:
When stored in a cool place (refrigerator), the stock solution remains stable for one month. The diluted investigational solutions must be used immediately.
For use with test kits:
Measuring range
in mg/l
102552
Spectroquant® Surfactants (anion.) Cell Test
0.05 - 2.00 MBAS