Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Analytical Quality Assurance, Standard for Tin
Reagents:
Cat.No. 170242  Tin standard solution Certipur®
Cat.No. 109057  Hydrochloric acid 1 mol/l Titripur®
Cat.No. 116754  Water for analysis
Preparation of a standard solution:
Transfer 30 ml of HCl 1 mol/l to a calibrated or conformity-checked 100-ml volumetric flask, add 10.0 ml (full pipette) of the tin standard solution, and make up to the mark with distilled water.
The standard solution prepared according to this procedure has a concentration of 100 mg/l tin.
Further investigational concentrations may be prepared from the standard solution by diluting accordingly with distilled water and HCl 1 mol/l. This is done in the following manner:
Transfer 1 ml of HCl 1 mol/l to a calibrated or conformity-checked 100-ml volumetric flask. Withdraw the desired aliquot from the tin standard solution 100 mg/l, add, make up to the mark with distilled water, and mix.
Stability:
The tin standard solution 100 mg/l remains stable for 30 minutes. The further diluted standard solutions (investigational concentrations) are not stabile and must be used immediately.
For use with test kit: Measuring range in mg/l
114622 Spectroquant® Tin Cell Test
 0.10 – 2.50 Sn