Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Analytical Quality Assurance, Standard for Total Hardness
Reagents:
Cat.No. 102121  Calcium nitrate tetrahydrate for analysis
Cat.No. 116754  Water for analysis
Preparation of a standard solution:
Dissolve 2.946 g of calcium nitrate tetrahydrate in a calibrated or conformity-checked 500-ml volumetric flask and make up to the mark with distilled water. The stock solution prepared according to this procedure has a concentration of 1000 mg/l calcium (corresponds to 175 °e).
Further investigational concentrations may be prepared from this stock solution by diluting accordingly.
Stability:
The stock solution of 1000 mg/l remains stable for one week. The diluted investigational solutions remain stable for one day.
For use with test kits:
Measuring range
110025
MQuantTM Total Hardness Test
<3 - >21 °d
<4 - >26 °e
<5 - >37 °f
110046
MQuantTM Total Hardness Test
>5 - >25 °d
>6 - >31 °e
>9 - >45 °f
117934
MQuantTM Total Hardness Test
<150 - >250 mg/l CaCO3
<1.5 - >2.5 mmol/l CaCO3<
<8.4 - >14 °d
<10.5 - >17.5 °e
<15 - >25 °f
108047
MColortestTM Total Hardness Test
1 - 100 mg/l CaCO3
0.1 - 5.6 °d
0.13 - 7 °e
0.18 - 10 °f
For use with test kits:
Measuring range
108039
MColortestTM Total Hardness Test
0.1 - 3.6 mmol/l CaCO3 (Ca)
0.2 - 20 °d
0.25 - 25 °e
0.36 - 35.6 °f
108312
MColortestTM Total Hardness Test
20 mg/l CaCO3
111104
MColortestTM Total Hardness Test
>1 °d
> 1.25 °e
>1.78 °f
> 7.1 mg/l Ca
116997
Reflectoquant® Total Hardness Test
0.1 -30.0 °d
0.1 - 37.6 °e
0.2 -53.5 °f
100961
Spectroquant® Total Hardness Cell Test
5 - 215 mg/l Ca
0.7 - 30.1 °d
0.9 - 37.6 °e
1.2 - 53.7 °f