Analytical Method: Copper (total) in steels (Others)
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114553  Spectroquant® Copper Cell Test
Cat.No. 114767  Spectroquant® Copper Test
Cat.No. 100317  Hydrochloric acid 37 % for analysis
Cat.No. 100456  Nitric acid 65 % for analysis
Cat.No. 116754  Water for analysis
Sample preparation:
In a glass beaker treat 100 mg (exactly weighed) of the sample with 7 ml of hydrochloric acid 37 % and 3 ml nitric acid 65 % and evaporate almost to dryness on the steam bath or water bath. The residue should be completely dry, free of acid, crystalline and of a yellowish or greenish color (not dark brown or black).
Dissolve the final residue in about 10 ml water for analysis and transfer to a 1000-ml volumetric flask, into which about 1 - 2 ml hydrochloric acid 37 % has been put.
Analysis:
Determine with the above mentioned test kits
Calculation:
Copper (total) content in %  =   analysis value in mg/l Cu x 100
weight of sample in mg