Analytical Method: Copper (total) in wine
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114553  Spectroquant® Copper Cell Test
Cat.No. 114767  Spectroquant® Copper Test
Cat.No. 110731  Sulfuric acid 95 - 97 % for analysis
Cat.No. 107209  Hydrogen peroxide 30 % (Perhydrol®) for analysis
Cat.No. 105590  Sodium hydroxide solution 32 % for analysis
Cat.No. 109531  MColorpHastTM pH-indicator strips pH 0 - 6
Cat.No. 116754  Water for analysis
Hints:
Samples of wine have to be treated with acid for determination. The various components of wine can cause an under-valuation, if no sample preparation is done.
Sample preparation:
In a 400-ml Erlenmeyer flask evaporate 100 ml of the sample to about 1 ml on the heating plate at low heat. Mix the residue with 10 ml sulfuric acid 95-97 % for analysis and heat until white SO3-smoke arises. After cooling add 2 ml Perhydrol®, mix and heat again on the heating plate. If the sample turns brown, again add 2 ml Perhydrol® after cooling and heat till the SO3 smoke arises. Afterwards the residue should be clear and colorless to light yellow. Finally evaporate the solution to a moist crystalline residue. After cooling dissolve the residue in 25 ml water for analysis and adjust to pH 3 - 4 with sodium hydroxide solution 32 % (pH-indicator strips pH 0 - 6). Transfer the solution into a 50-ml volumetric flask, fill up to the mark with water for analysis and mix.
Analysis:
Determine with the above mentioned test kits
Calculation:
Copper (total) content in mg/l Cu =   analysis value in mg/l Cu
2