TLC Analysis Analgesics and Preservatives on HPTLC Silica Gel 60 RP-18 WF254s (analyte set 4)
TLC Analysis Analgesics and Preservatives on HPTLC Silica Gel 60 RP-18 WF254s (analyte set 4)
Conditions
sample 0.2 µL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 248 nm
plate HPTLC pre-coated plate RP-18 wF 254s (1.13124.0001)
chamber normal chamber without chamber saturation
eluent [A] acetonitrile; [B] water; (40:60, A:B)
migration distance 4.5 cm
Description
Categories Analytical Chromatography, TLC Applications, analgesics
Featured Industry Pharmaceutical (small molecule)
General description TLC Plate Information: 1.13124.0001
suitability application for TLC