TLC Analysis of Alanine Oligomers on HPTLC Silica gel 60 RP-18 F254s
TLC Analysis of Alanine Oligomers on HPTLC Silica gel 60 RP-18 F254s
Conditions
detector ninhydrin spray reagent, heated to 110 °C for 15 min
eluent [A] acetonitrile; [B] ethanol; [C] water; (90:5:10, A:B:C) containing 0.1 M chloride
plate HPTLC pre-coated plate RP-18 F 254s (1.15389.0001, 1.15423.0001)
chamber normal chamber without chamber saturation
migration distance 12 cm
Description
Categories Analytical Chromatography, TLC Applications, peptides
Featured Industry Life Science and Biopharma
General description TLC Plate Information: 1.15389.0001
suitability application for TLC