TLC Analysis of Aromatic Carboxylic Acids on HPTLC Silica Gel 60 RP-18 WF254s
TLC Analysis of Aromatic Carboxylic Acids on HPTLC Silica Gel 60 RP-18 WF254s
Conditions
sample 0.2 µL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
plate HPTLC pre-coated plate RP-18 F 254s (1.13124.0001)
migration distance 5 cm
chamber normal chamber without chamber saturation
eluent [A] methanol; [B] water; (60:40, A:B) with addition
of 0.1 M tetramethylammoniumbromide
Description
Categories Analytical Chromatography, TLC Applications, organic acids
Featured Industry Chemicals and Industrial Polymers
General description TLC Plate Information: 1.13724.0001
suitability application for TLC