TLC Analysis of Aromatic Carboxylic Acids on HPTLC Silica Gel 60 RP-8 F254s
TLC Analysis of Aromatic Carboxylic Acids on HPTLC Silica Gel 60 RP-8 F254s
Conditions
sample 0.2 µL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
migration distance 5 cm
plate HPTLC pre-coated plate RP-8 F 254s (1.13725.0001)
chamber normal chamber without chamber saturation
eluent [A] methanol; [B] water; (60:40, A:B) containing 0.1 M TEABr
Description
Categories Analytical Chromatography, TLC Applications, carboxylic acids
Featured Industry Chemicals and Industrial Polymers
suitability application for TLC