TLC Analysis of Barbiturates and Methaqualone on HPTLC Silica Gel 60 RP-8 F254s
TLC Analysis of Barbiturates and Methaqualone on HPTLC Silica Gel 60 RP-8 F254s
Conditions
sample 0.3 µl
detector in situ-evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
test parameters eluent: [A] methanol; [B] water; (80:20, A:B)
test parameters plate: TLC pre-coated plate RP-8 F 254s (1.15388.0001, 1.15424.0001)
test parameters chamber: normal chamber without chamber saturation
test parameters migration distance: 7 cm
Description
Categories Analytical Chromatography, TLC Applications, drugs of abuse
Featured Industry Clinical
Forensics and Toxicology
Pharmaceutical (small molecule)
suitability application for TLC