TLC Analysis of Benzoic Acids on HPTLC Silica Gel 60 RP-18 WF254s
TLC Analysis of Benzoic Acids on HPTLC Silica Gel 60 RP-18 WF254s
Conditions
sample 0.3 µL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
migration distance 7 cm
plate HPTLC pre-coated plate RP-18 wF 254s (1.13124.0001)
chamber normal chamber without chamber saturation
eluent [A] acetone; [B] water; (40:60, A:B)
Description
Categories Analytical Chromatography, TLC Applications, organic acids
Featured Industry Chemicals and Industrial Polymers
General description TLC Plate Information: 1.13124.0001
suitability application for TLC