TLC Analysis of Chlorophenols on HPTLC Silica gel 60 RP-18 F254s
TLC Analysis of Chlorophenols on HPTLC Silica gel 60 RP-18 F254s
Conditions
detector in-situ evaluation with TLC/HPTLC scanner (Camag) UV 265 nm
migration distance 7 cm
chamber normal chamber without chamber saturation
eluent [A] acetonitrile; [B] water; (80:20, A:B)
plate HPTLC pre-coated plate RP-18 WF 254s (1.13724.0001)
Description
Categories Analytical Chromatography, TLC Applications, phenols
Featured Industry Chemicals and Industrial Polymers
General description TLC Plate Information: 1.13724.0001
suitability application for TLC