TLC Analysis of Dipeptides on HPTLC Silica Gel 60 RP-18 F254s
TLC Analysis of Dipeptides on HPTLC Silica Gel 60 RP-18 F254s
Conditions
sample 0.3 µL
detector spray reagent: ninhydrin, heated to 110 °C for 10 min
plate TLC pre-coated plate RP-18 F 254s (1.15389, alt. P/N 1.15423)
migration distance 8 cm
chamber normal chamber without chamber saturation
eluent [A] acetonitrile; [B} water; (40:60, A;B) containing 0.1 M lithium chloride
Description
Categories Analytical Chromatography, TLC Applications, peptides
Featured Industry Life Science and Biopharma
General description TLC Plate Information: 1.15389.0001
suitability application for TLC