TLC Analysis of Drugs on HPTLC Silica Gel 60 RP-18 F254s
TLC Analysis of Drugs on HPTLC Silica Gel 60 RP-18 F254s
Conditions
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
chamber normal chamber without chamber saturation
plate TLC pre-coated plate RP-18 F 254s (1.15389.0001)
eluent [A] acetone; [B] water; (50:50, A:B) containing 0.1 M tetramethylammoniumbromide
Description
Categories Analytical Chromatography, TLC Applications, drugs of abuse, drugs, miscellaneous
Featured Industry Pharmaceutical (small molecule)
General description TLC Plate Information: 1.15389.0001
suitability application for TLC