TLC Analysis of Ingredients in Analgesic Agents on HPTLC Silica Gel 60 RP-18 wF 254s with Concentrating Zone
TLC Analysis of Ingredients in Analgesic Agents on HPTLC Silica Gel 60 RP-18 wF 254s with Concentrating Zone
Conditions
sample 5 µl
detector dipped in dichlorofluorescein (0.2% in ethanol) (in situ evaluation with TLC/HPTLC
scanner (Camag) UV 265 nm)
test parameters chamber: normal chamber without chamber saturation
test parameters migration distance: 7 cm
test parameters eluent: [A] acetonitrile; [B] water; (40:60, A:B) containing 0.1 M ammonium bromide
test parameters plate: HPTLC pre-coated plate RP-18 wF 254s with concentrating zone (1.13124.0001)
Description
Featured Industry Life Science and Biopharma
suitability application for TLC