TLC Analysis of Ingredients in Expectorants on HPTLC Silica Gel 60 RP-18 WF254s
TLC Analysis of Ingredients in Expectorants on HPTLC Silica Gel 60 RP-18 WF254s
Conditions
sample 0.2 µL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 248 nm
eluent [A] acetonitrile; [B] water; (40:60, A:B) containing 0.1 M TMABr
plate HPTLC pre-coated plate RP-18 wF 254s with concentrating zone
(1.13124.0001&Brand=MM">1.13124.0001)
chamber normal chamber without chamber saturation
migration distance 7 cm
Description
Categories Analytical Chromatography, TLC Applications, peptides
Featured Industry Life Science and Biopharma
suitability application for TLC