TLC Analysis of Mycotoxins on HPTLC Silica Gel 60 RP-8 WF254s
TLC Analysis of Mycotoxins on HPTLC Silica Gel 60 RP-8 WF254s
Conditions
sample 0.3 µl
detector in situ-evaluation with TLC/HPTLC scanner (Camag) UV 248 nm
test parameters plate: TLC pre-coated plate RP-8 F 254s (1.15388.0001, 1.15424.0001)
test parameters migration distance: 8 cm
test parameters chamber: normal chamber without chamber saturation
test parameters eluent: [A] acetonitrile; [B] buffer solution pH 2.00; (50:50, A:B)
Description
Categories Analytical Chromatography, TLC Applications, mycotoxins
Featured Industry Food and Beverages
General description TLC Plate Information: 1.15388.0001
suitability application for TLC