TLC Analysis of Nucleoside Monophosphates on HPTLC Pre-coated Plate RP-18 WF 254s with Concentrating Zone
TLC Analysis of Nucleoside Monophosphates on HPTLC Pre-coated Plate RP-18 WF 254s with Concentrating Zone
Conditions
sample 5 µL, 0.01%
detector In-situ evaluation with TLC/HPTLC scanner (Camag) UV 260 nm
test parameters plate: HPTLC pre-coated plate RP-18 WF 254s with concentrating zone (1.13124.0001)
test parameters chamber: normal chamber without chamber saturation
test parameters eluent: water containing 0.2M 1-butanesulfonic acid sodium salt
test parameters migration distance: 5 cm
Description
suitability application for TLC