TLC Analysis of Opioid Analgesics on HPTLC Silica Gel 60 RP-18 WF254s
TLC Analysis of Opioid Analgesics on HPTLC Silica Gel 60 RP-18 WF254s
Conditions
sample 0.3 µL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
eluent [A] methanol; [B] water; (30:70, A:B) containing 0.1 M TMABr
plate HPTLC pre-coated plate RP-18 WF 254s (1.13124.0001)
migration distance 5 cm
chamber normal chamber without chamber saturation
Description
Categories Analytical Chromatography, TLC Applications, opiates, analgesics
Featured Industry Pharmaceutical (small molecule)
General description TLC Plate Information: 1.13124.0001
suitability application for TLC