TLC Analysis of Tripeptides by Ion Pair Chromatography on HPTLC Silica Gel 60 RP-18 F254s
TLC Analysis of Tripeptides by Ion Pair Chromatography on HPTLC Silica Gel 60 RP-18 F254s
Conditions
sample 0.7 µL
detector spray reagent ninhydrin, heated to 120 °C for 10 min
plate HPTLC pre-coated plate RP-18 F 254s (1.15389.0001, 1.15423.0001)
eluent [A] acetonitrile; [B] 1 N acetic acid; (98:8, A:B) containing
0.05 M octanesulfonic acid sodium salt
chamber normal chamber without chamber saturation
migration distance 12 cm
Description
Categories Analytical Chromatography, TLC Applications, General TLC Applications
Featured Industry Life Science and Biopharma
General description TLC Plate Information: 1.15389.0001
suitability application for TLC