TLC Analysis of Tripeptides on HPTLC Silica Gel 60 RP-18 F254s (analyte set 2)
TLC Analysis of Tripeptides on HPTLC Silica Gel 60 RP-18 F254s (analyte set 2)
Conditions
detector spray reagent: ninhydrin, heated to 110 °C for 20 min
migration distance 7 cm
eluent [A] acetonitrile; [B] buffer solution, pH 3.00; (90:10, A:B) containing
0.1 M lithium chloride
chamber normal chamber without chamber saturation
plate TLC pre-coated plate RP-18 F 254s (1.13724.0001)
Description
Categories Analytical Chromatography, TLC Applications, peptides
Featured Industry Life Science and Biopharma
General description TLC Plate Information: 1.13724.0001
suitability application for TLC