Aquastar® Water Standard 5 mg/ml
This liquid water standard is based on a long-chain alcohol. Under normal conditions the adjusted water content is maintained after opening the bottle. This water standard 5 mg/ml can be used for daily titer control, but is not recommended for exact titer determinations.
Materials
If you have trouble finding the products you're looking for, please contact our customer support for assistance.