Phytochemical Snap-N-Shoot® Reference Solutions
Sigma-Aldrich's Cerilliant brand offers Snap-N-Shoot® reference solutions and mixes for several botanicals including Turmeric, Ephedra, Ginkgo biloba, Ginseng, and Green tea. Several of these reference solutions include multi-component mixes of the most common phytochemicals found in each botanical. Certified concentration values for the components in each reference solution improve laboratory efficiency from less time weighing powders or extracts and preparing stock solutions to rerunning samples or remaking stocks due to possible differences in raw material purity factors. The convenient 1 mL unit size minimizes waste and additional storage of sensitive phytochemicals in solution under non-inert environments. Rigorous testing, including accelerated and real time studies, ensures solution accuracy and shelf life.
Cerilliant certified reference solutions are prepared and certified to the highest industry standards including ISO Guide 34 and ISO/IEC 17025. Each Snap-N-Shoot® solution is supported by a comprehensive Certificate of Analysis (COA) which provides all analytical data and uncertainty and traceability information to support regulatory requirements. Our COA includes analytical data on homogeneity and concentration certification as well as accuracy of concentration by comparison to an independently prepared calibration curve.
Certified Snap-N-Shoot® reference solutions of caffeine and other common dietary supplement adulterants such as amphetamines and erectile-dysfunction drugs are also available.