Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
TISSUE-TROL™ Control Slides
Histology control slides are essential in the support of formal quality assurance programs. Our TISSUE-TROL™ slides are comprised of paraffin embedded tissue sections containing known characteristics and are used for monitoring staining performance in pathology laboratories.
Argentaffin from Human Intestine
Argentaffin TISSUE-TROL™ Control SlidesArgentaffin TISSUE-TROL Control Slides are paraffin embedded sections of human intestine containing argentaffin granules.
Each box contains 24 slides plus one reference slide stained using Fontana-Masson Stain Kit (Product No. HT200).
Melanin from Human Skin
Melanin TISSUE-TROL™ Control SlidesMelanin TISSUE-TROL™ Control Slides are paraffin embedded sections of human skin containing melanin.
Each box contains 24 slides plus one reference slide stained using Fontana-Masson Stain Kit (Product No. HT200).