Extract-N-Amp™ Seed PCR Kit
Description
Extract-N-Amp Seed Flow Chart
The Extract-N-Amp Seed PCR Kits contain all the reagents necessary to rapidly extract genomic DNA from seeds and amplify targets of interest by PCR (Figure 1).
  • Novel – single-step extraction of seed genomic DNA to PCR
  • Fast – seed to PCR in 15 minutes
  • Convenient – no organic extraction, column purification or precipitation of DNA
The Extract-N-Amp Seed PCR Kit offers a novel extraction system that eliminates the need for DNA purification, organic extraction, centrifugation, heating, filtration or alcohol precipitation. The product includes a specially formulated hot start PCR ReadyMix™ for amplification directly from the extract. Extract-N-Amp products are Sigma Advanced Technology certified.
Procedure
Genomic DNA is extracted from ground seed material by incubation in a mixture of Extraction Solution and Seed Preparation Solution at 55 °C for 10 minutes. After the extraction is stopped by incubation at 95 °C for 3 minutes, an equal volume of Neutralization Solution B is added and the extract is ready for PCR. An aliquot of the extract is then added directly to the optimized PCR mix supplied.
Application
Perfect for genotyping.
Beneficial Features
Starting Material 1 seed
Speed Extraction of seed genomic DNA for PCR in less than 15 minutes
Versatile Extract and amplify target DNA sequences from a variety of plant species (Figure 1)
High Specificity Hot Start antibody for highly specific PCR amplification of genomic DNA
Safe No phenol/chloroform or other hazardous materials
Pick the formulation that is right for you
The Extract-N-Amp Tissue PCR Kits include a specially formulated hot start PCR ReadyMix for amplification directly from the extract.
The PCR ReadyMix comes in two formulations:
  • REDExtract-N-Amp PCR Ready Mix – contains an inert dye that acts as a tracking dye and allows for convenient loading of PCR reactions onto agarose gels for analysis.
  • Extract-N-Amp PCR Ready Mix – no dye contained.
Ordering Information
Extract-N-Amp Seed PCR Kits
Product No. Name Number of Extractions Number of Amplifications Technical Manual
XNAS2 Extract-N-Amp Seed PCR Kit 100 100 PDF
REDExtract-N-Amp Seed PCR Kits
Product No. Name Number of Extractions Number of Amplifications Technical Manual
XNASS REDExtract-N-Amp Seed PCR Kit
(contains REDTaq)
10 10 PDF
XNAS REDExtract-N-Amp Seed PCR Kit
(contains REDTaq)
100 100 PDF
XNASR REDExtract-N-Amp Seed PCR Kit
(contains REDTaq)
1000 1000 PDF
Kit Contents
Extract-N-Amp Kit
  • Seed Preparation Solution
  • Extraction Solution
  • Neutralization Solution B
  • Extract-N-Amp PCR Ready Mix
REDExtract-N-Amp Kit
  • Seed Preparation Solution
  • Extraction Solution
  • Neutralization Solution B
  • REDExtract-N-Amp PCR Ready Mix
Sample Data
Multiplex PCR analysis of Extract-N-Amp Seed PCR Kit extracts from various seed types
Figure 1. Genomic DNA was extracted from the seeds using the protocol as described in the Extract-N-Amp Seed Technical Bulletin. All extracts were then amplified using the specially formulated JumpStart™ PCR mix and PCR primers multiplexed for both a universal chloroplast gene (~400-700 bp) and the acetylcoenzyme A carboxylase gene specific to wheat (964 bp).
Stability of Soybean Extracts at 4 °C
Figure 2. Soybean seeds were extracted according to the procedure in the Technical Bulletin for the Extract-N-Amp Seed PCR Kit. 4 µL aliquots were analyzed immediately by quantitative PCR with SYBR Green detection on an ABI Prism® 7700. DNA standards for quantitative PCR were purified DNA prepared from soybean seeds using the GenElute Plant Genomic DNA Kit (Product No. G2N70) and stored as single use aliquots at -20 °C. The soybean seed extracts were stored at 4 °C (recommended storage). Quantitative PCR was repeated after 2.5 and 5 weeks and then 2 and 6 months for the 4 °C samples. These results show that the extracts will be stable for at least 6 months at the recommended storage temperature of 4 °C.
REDTaq allows for convenient loading onto agarose gels
Figure 3. Samples are loaded directly on an agarose gel after PCR amplification with RedTaq DNA Polymerase, obviating the need or addition of loading buffers and/or tracking dyes. The RedTaq dye approximately co-migrates with a 125 bp duplex.