Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Acetylcholine Synthesis and Metabolism
Acetylcholine is synthesized from acetyl coenzyme A and choline by the enzyme choline acetyltransferase. In the nervous system, this enzyme is thought to exist primarily in the nerve terminal cytoplasm. Coenzyme A is synthesized in mitochondria and accesses choline acetyltransferase following transport across the mitochondrial membrane into the cytoplasm. In addition to its synthesis in the liver, choline employed in acetylcholine production is derived from dietary sources. There is a carrier system in capillary endothelial cells that is responsible for transport of choline, in its free and phospholipid forms, into the brain. Hydrolysis of choline-containing phospholipids may also liberate choline that is used in acetylcholine synthesis. As choline acetyltransferase is not saturated by concentrations of acetyl coenzyme A and choline that are estimated to be present in the nerve terminal, it appears that the rate of acetylcholine synthesis is dependent on precursor availability. Enzyme activity is also regulated by product inhibition; by binding at an allosteric site on choline acetyltransferase, acetylcholine inhibits its activity. In addition, mass action and neuronal activity influence the rate of acetylcholine formation. Short-term regulation of enzyme activity is partly achieved by phosphorylation induced by protein kinases. There are no very specific and potent inhibitors of the enzyme and it should be noted that pharmacological blockade of this step (e.g. with naphthylvinylpyridine) in the life-cycle of acetylcholine produces a less profound effect on the transmitter than does inhibition of choline transport.
A specific low-affinity acetylcholine transporter is responsible for uptake of the transmitter from the cytoplasm into vesicles. The genes for choline acetyltransferase and the vesicular acetylcholine transporter are organized in a single gene locus, and transcription of the two genes is typically co-regulated. (±)-Vesamicol is a selective inhibitor of this transporter, with L-(–)-vesamicol being more potent than D-(+)-vesamicol. Once packaged in vesicles, acetylcholine is subject to stimulus-induced release by exocytosis. Several powerful toxins impact on acetylcholine release, notably botulinum toxin which inhibits its release.
Neuronal acetylcholinesterase very rapidly inactivates the majority of acetylcholine released in brain, although butyrylcholinesterase contained in glial cells may hydrolyze a small proportion of acetylcholine in the synapse. In the periphery, acetylcholinesterase is present in muscle that receives cholinergic innervation, while butyryl-cholinesterase is more widely distributed. A number of reversible (e.g. physostigmine, BW284C51) and irreversible (e.g. iso-OMPA) inhibitors of acetylcholinesterase are known, and these drugs have the effect of prolonging the synaptic effects of acetylcholine. Second generation reversible anticholinesterases such as donepezil, rivastigmine (ENA 713), eptastigmine, and galantamine (galanthamine) are being employed as treatments for Alzheimer’s disease. Some second generation cholinesterases have been withdrawn from clinical use because of unacceptable side effects (e.g, tacrine, metrifonate). Irreversible acetylcholinesterase inhibitors are used as insecticides and chemical warfare agents. Choline, which is liberated from acetylcholine by acetylcholinesterase, is taken back up into cholinergic terminals by a high-affinity transporter, and reused in transmitter synthesis. Hemicholinium-3 potently and reversibly inhibits choline transport, and this results in a profound decrease in acetylcholine formation. Unlike hemicholinium-3, A-4 (a bis 4-methylpiperidine analog of HC-3), is active following peripheral administration. Nitrogen mustard analogs of choline are potent irreversible inhibitors of high-affinity choline uptake.
The Table below contains accepted modulators and additional information. For a list of additional products, see the "Similar Products" section below.
Inhibitors   Uptake Compound Enzyme Co-Factors Inhibitors
Hemicholinium-3 (H108)
Choline mustard aziridinium
A-4
←←←
→→→→
←←←←
Choline (C1789, C7017)
↓→→→→→→→→→→→→→→
Acetylcholine (A2661, A6625)
↓→→→→→→→→→→→→→→
Choline (C1789, C7017) + Acetic acid (320099)
Choline acetyltransferase
Acetylcholinesterase (C1682)
Acetyl CoA (A2056, A2181)
Naphthylvinylpyridine
Eserine (E8375)
N-Methylphysostigmine (M100)
Neostigmine (N2001)
Ambenonium dichloride
Tacrine (A3773)
Edrophonium chloride
BW 284C51 (A9013)
ISO-OMPA (T1505)
SDZ ENA 173
9-Amino-1,2,3,4-tetrahydroacridine bis 1,7-heptylene (A256)
Galanthamine (G1660)
Inhibitors Uptake Compound Enzyme Co-Factors Inhibitors
Hemicholinium-3 (H108)
Choline mustard aziridinium
A-4
Choline (C1789, C7017)
│––––––––––––––––––––––––––→
Acetylcholine (A2661, A6625)
│––––––––––––––––––––––––––→
Choline (C1789, C7017) + Acetic acid (320099)
Choline acetyltransferase
Acetylcholinesterase (C1682)
Acetyl CoA (A2056, A2181)
Naphthylvinylpyridine
Eserine (E8375)
N-Methylphysostigmine (M100)
Neostigmine (N2001)
Ambenonium dichloride
Tacrine (A3773)
Edrophonium chloride
BW 284C51 (A9013)
ISO-OMPA (T1505)
SDZ ENA 173
9-Amino-1,2,3,4-tetrahydroacridine bis 1,7-heptylene (A256)
Galanthamine (G1660)
Abbreviations
ISO-OMPA: Tetraisopropyl pyrophosphoramide
Similar Products
References